Learn More
Reitenspiess™ Test Tube Brush
Product Code. 11351265
Change view
Click to view available options
Length (Metric):
100 mm
190 mm
230 mm
Diameter (Metric) Bristle:
16 mm
26 mm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Bristle unitSize
11351265 230 mm 26 mm Each
11361265 190 mm 16 mm Each
11371265 - - Each
11327494 - - Each
11337494 100 mm - Each
5 options
Product Code. 11351265
Brand: Reitenspiess™ 12240301
Only null left
Add to basket
Add to basket
Use a Reitenspiess™ Test Tube Brush for cleaning, disinfecting and sterilizing test tubes as necessary in the laboratory. .
  • Natural bristle
  • Various lengths available
  • Various diameters
Specifications
Product Type Test Tube Brush
Diameter (Metric) Bristle 26 mm
Length (Metric) 230 mm
Quantity 1 Ea.