Learn More
IKA™ Test Tube Attachment for IKA Platform Shaker
Product Code. 10728971 Shop All IKA Products
Change view
Click to view available options
Holds:
Tubes
Unit Size:
Each
Product Code. Holds unitSize
10728971 Tubes Each
10490243 - Each
10130921 Tubes Each
3 options
Product Code. 10728971
Brand: IKA™ L001840
Only null left
Add to basket
Add to basket
Attachment with customized array for holding reagent glasses on a platform shaker.
  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
TRUSTED_SUSTAINABILITY
Specifications
Holds Tubes
Tube Size 10 mm
Product Type Test Tube Adapter Attachment
Description Test tube adapter attachment
Model Model MS 1.31
For Use With (Equipment) Universal attachment, shaker
No. of Positions 14