Learn More
IKA™ Test Tube Attachment for IKA Platform Shaker
Shop All IKA Products
Click to view available options
Holds:
Tubes
Unit Size:
Each
Description
Attachment with customized array for holding reagent glasses on a platform shaker.
- Requires VG 3.3 universal attachment
- Facilitates multi-tube vortexing
Specifications
Specifications
| Holds | Tubes |
| Tube Size | 12 mm |
| Product Type | Test Tube Adapter Attachment |
| Description | Test tube adapter attachment |
| Model | Model MS 1.32 |
| For Use With (Equipment) | Universal attachment, shaker |
| No. of Positions | 6 |