Learn More
Tesevatinib, MedChemExpress
Product Code. 16475638
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16475638
Brand: Medchem Express HY13314
MedChemExpress Tesevatinib (XL-647; EXEL-7647; KD-019) is an orally available, multi-target tyrosine kinase inhibitor; inhibits EGFR, ErbB2, KDR, Flt4 and EphB4 kinase with IC50s of 0.3, 16, 1.5, 8.7, and 1.4 nM.
Tesevatinib (XL-647
Specifications
Chemical Name or Material Tesevatinib
CAS 781613-23-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H25Cl2FN4O2
Quantity 10 mg
Synonym XL-647 EXEL-7647 KD-019
Solubility Information DMSO : 100 mg/mL (203.50 mM; Need ultrasonic)
SMILES [H][C@@]12[C@@](C[C@@H](COC3=CC4=C(C(NC5=C(F)C(Cl)=C(Cl)C=C5)=NC=N4)C=C3OC)C2)([H])CN(C)C1
Molecular Weight (g/mol) 491.39
Formula Weight 491.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.