Learn More
tert-Butylphosphonic acid, 98%
Product Code. 10301094 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. 10301094
Brand: Thermo Scientific Acros 321520050
Request Bulk or Custom Format
CAS: 4923-84-6 | C4H9O3P | 136.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4923-84-6
Molecular Formula C4H9O3P
Molecular Weight (g/mol) 136.09
MDL Number MFCD00012296
InChI Key OGDSVONAYZTTDA-UHFFFAOYSA-L
Synonym phosphonic acid, 1,1-dimethylethyl, acmc-20ajka, t-butylphosphonic acid, tert-butylphosphonic acid, 2-methylprop-2-ylphosphonic acid, phosphonicacid, 1,1-dimethylethyl
PubChem CID 312546
IUPAC Name tert-butylphosphonic acid
SMILES CC(C)(C)P([O-])([O-])=O
Specifications
Melting Point 188.0°C to 192.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CP(O)(OH)2
Quantity 5 g
Formula Weight 138.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylphosphonic acid