missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene, 99%

Product Code. 10311564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
Product Code. Quantity Packaging unitSize
10311564 2.5 L Glass bottle 2.5 L
10204040 500 mL Glass bottle 500mL
10346170 1 L Glass bottle 1 L
3 options
This item is not returnable. View return policy

Product Code. 10311564

Brand: Thermo Scientific Acros 107870025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-06-6 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Melting Point -58.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 169.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C(CH3)3
Refractive Index 1.492
Quantity 2.5 L
Beilstein 05, 415
Merck Index 15, 1553
Specific Gravity 0.86
Solubility Information Solubility in water: 0.03g/L water (20°C). Other solubilities: miscible with alcohol,ether and benzene
Viscosity 1.15 mPa.s (20°C)
Formula Weight 134.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butylbenzene
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.