Learn More
tert-Butylamine, Sigma-Aldrich™
Product Code. 15898570
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
35 kg
Unit Size:
1 L
100mL
35kg
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898570 500 mL 500mL
15888570 100 mL 100mL
15808580 1 L 1 L
15818580 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15898570 Supplier Sigma Aldrich Supplier No. 8015460500
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-64-9
Specifications
Melting Point -72°C
Density 0.69 g/cm3 at 20°C
Boiling Point 44°C
Molecular Formula (CH3)3CNH2
MDL Number MFCD00008050
Quantity 500 mL
Synonym tert-Butylamine, 2-Amino-2-methylpropane, tert-Butylamine
Molecular Weight (g/mol) 73.14
Grade Research