Learn More
tert-Butyl (Ethyl-d5)(isopropyl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl (Ethyl-d5)(isopropyl)carbamate |
| Molecular Formula | C10 D5 H16 N O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C10H21NO2/c1-7-11(8(2)3)9(12)13-10(4,5)6/h8H,7H2,1-6H3/i1D3,7D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])N(C(C)C)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-(1,1,2,2,2-pentadeuterioethyl)-N-propan-2-ylcarbamate |
| Molecular Weight (g/mol) | 192.31 |
| Formula Weight | 192.189 |