Learn More
tert-Butyl carbazate, 99%
Product Code. p-79265 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. 10692002
Brand: Thermo Scientific Acros 107880250
Request Bulk or Custom Format
CAS: 870-46-2 | C5H12N2O2 | 132.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 870-46-2
Molecular Formula C5H12N2O2
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007593
InChI Key DKACXUFSLUYRFU-UHFFFAOYSA-N
Synonym tert-butyl carbazate, tert-butyl hydrazinecarboxylate, t-butyl carbazate, hydrazinecarboxylic acid, 1,1-dimethylethyl ester, t-butylcarbazate, tert-butoxycarbonyl hydrazide, boc-hydrazide, boc-hydrazine, tert-butoxycarbonyl hydrazine, carbazic acid tert-butyl ester
PubChem CID 70091
IUPAC Name tert-butyl N-aminocarbamate
SMILES CC(C)(C)OC(=O)NN
Specifications
Melting Point 37°C to 41°C
Color White to Yellow
Density 1.0200g/mL
Boiling Point 63°C to 65°C (0.1 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NNHCO2C(CH3)3
Quantity 25 g
Fieser 01,85; 02,46; 11,88
Specific Gravity 1.02
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 132.16
Percent Purity 99%
Physical Form Crystalline Lumps
Chemical Name or Material tert-Butyl carbazate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl carbazate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.