missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl carbamate, 98+%

Product Code. p-7043328
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11349963 1 g 1g
11359963 5 g 5g
11369963 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11349963

Brand: Thermo Scientific Alfa Aesar L00259.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4248-19-5 | C5H11NO2 | 117.148 g/mol

tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

Solubility
Soluble in methylene chloride, chloroform, and alcohols. Slightly soluble in petroleum ether and water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4248-19-5
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00007962
InChI Key LFKDJXLFVYVEFG-UHFFFAOYSA-N
Synonym t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
PubChem CID 77922
IUPAC Name tert-butyl carbamate
SMILES CC(C)(C)OC(=O)N

Specifications

Melting Point 106°C to 110°C
Beilstein 1744500
Solubility Information Soluble in methylene chloride,chloroform,and alcohols. Slightly soluble in petroleum ether and water.
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥98%
Chemical Name or Material tert-Butyl carbamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.