Learn More
tert-Butanol, Supelco™
Product Code. 15896720
Change view
Click to view available options
Quantity:
1 L
25 L
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896720 1 L 1 L
15841741 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896720 Supplier Supelco Supplier No. 8222641000
 Restricted Item
Estimated Shipment: 16-11-2026
EMPLURA™
EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-65-0
Specifications
Melting Point 24°C to 25°C
Density 0.78 g/cm3 at 20°C
Quantity 1 L
Molecular Formula (CH3)3COH
MDL Number MFCD00004464
Synonym tert-Butanol, tert-Butyl alcohol, 2-Methyl-2-propanol, Trimethylcarbinol, tert-Butanol
Boiling Point 81°C to 83°C/1013 hPa
Molecular Weight (g/mol) 74.12
Grade Research
Chemical Name or Material Tert-Butanol