Learn More
Thermo Scientific™ Terry Tool Tubing Cutters
Product Code. 10403685 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
1/16 in. Stainless Steel Tubing
1/8 in. Stainless Steel Tubing
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10403685 1/16 in. Stainless Steel Tubing Each
10260765 1/8 in. Stainless Steel Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10403685 Supplier Thermo Scientific™ Supplier No. 60182509
Estimated Shipment: 29-12-2026
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Cutter
Description Cuts 0.0625 stainless steel tubing
For Use With (Equipment) 1/16 in. Stainless Steel Tubing