Learn More
Thermo Scientific™ Terry Tool Tubing Cutters
Product Code. 10260765 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
1/16 in. Stainless Steel Tubing
1/8 in. Stainless Steel Tubing
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10260765 1/8 in. Stainless Steel Tubing Each
10403685 1/16 in. Stainless Steel Tubing Each
2 options
Product Code. 10260765
Brand: Thermo Scientific™ 60182510
Only null left
Add to basket
Add to basket
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Cutter
Description Cuts 0.125 stainless steel tubing
For Use With (Equipment) 1/8 in. Stainless Steel Tubing