Learn More
3M™ Nomad™ Terra Medium Traffic Entrance Matting 6050
Product Code. 17178730
Change view
Click to view available options
Dimensions (L x W):
152 x 91.4 cm
610 x 91.4 cm
180 x 122 cm
610 x 122 cm
Color:
Black
Blue
Brown
Gray
Red
Unit Size:
1 roll
Each
Product Code. Dimensions (L x W) Color unitSize
17178730 610 x 91.4 cm Black 1 roll
17158720 152 x 91.4 cm Gray 1 roll
17198720 180 x 122 cm Gray 1 roll
17168720 610 x 122 cm Red 1 roll
17158730 610 x 91.4 cm Blue Each
17168730 610 x 91.4 cm Brown 1 roll
17188720 610 x 91.4 cm Gray Each
17198730 610 x 122 cm Black 1 roll
17108730 610 x 122 cm Blue 1 roll
17148730 610 x 122 cm Brown 1 roll
17178740 610 x 122 cm Gray 1 roll
11 options
Product Code. 17178730
Brand: 3M™ Nomad™ 7100116713
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This backed vinyl matting is suitable for indoor, medium traffic applications. Resilient vinyl loop construction traps, holds, and hides dirt/sand so surfaces stay neat and clean.
  • First step mat for efficient dirt removal
  • Suitable for medium traffic volumes
  • Collects and prevents dirt from being tracked into the building
  • Holds and hides dirt & sand, so the surface stays neat and clean
  • Foam backing reduces creeping and, together with the coils, provides additional cushioning for increased comfort
  • Easy installation
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 610 x 91.4 cm
For Use With (Application) Medium traffic areas
Thickness (Metric) 11 mm
Color Black
Length (Metric) 610 cm
Material Vinyl
Type Entrance Mat
Width (Metric) 91.4 cm