Learn More
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8100
Entrance coiled vinyl scraper matting designed for heavy traffic applications. The construction allows dirt and moisture to flow through the matting so it is not retracked into the building. Grease and oil resistant.
Brand: 3M™ Nomad™ 7100116630
Additional Details : Weight : 26.76200kg
Description
- Unbacked scraper matting designed for high traffic applications
- High quality vinyl offers excellent crush resistance
- Unbacked, open construction of the mat allows water to drain and soil to pass through, out of sight
- Easily vacuum out trapped dirt
Specifications
610 x 91.4 cm | |
Heavy traffic areas | |
17 mm | |
91.4 cm |
Brown | |
Nomad™ Terra Dirt Control Mats | |
Elastic Vinyl | |
610 cm |