Learn More
Terpineol (>90%), TRC
Product Code. 30138470
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138470 5 g 5g
30137889 25 g 25g
30005266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138470 Supplier Toronto Research Chemicals Supplier No. TRCT1166255G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8000-41-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
Synonym Terpineol, Alpha Jax, Terpineol 318, Terpineol C, Terpineol LW
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)cyclohexan-1-ol--1-methyl-4-(propan-2-ylidene)cyclohexan-1-ol--2-(4-methylcyclohex-3-en-1-yl)propan-2-ol (1/1/1)
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2
Specifications
Quantity 5 g
InChI Formula InChI=1S/3C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;2*1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;11H,4-7H2,1-3H3;9,11H,1,4-7H2,2-3H3
Formula Weight 154.14
Chemical Name or Material Terpineol