Learn More
Terlipressin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Terlipressin is a vasopressin analogue with potent vasoactive properties. Terlipressin is a highly selective vasopressin V1 receptor agonist that reduces the splanchnic blood flow and portal pressure and controls acute variceal bleeding. Terlipressin exerts anti-inflammatory and anti-oxidative effects. Terlipressin has the potential for hepatorenal syndrome and norepinephrine-resistant septic shock research.
Specifications
Specifications
| Chemical Name or Material | Terlipressin |
| CAS | 14636-12-5 |
| Purity Grade Notes | Research |
| Molecular Formula | C52H74N16O15S2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (81.48 mM; Need ultrasonic) |
| SMILES | O=C([C@H](CSSC[C@@H](C(N[C@H](C1=O)CC2=CC=C(O)C=C2)=O)NC(CNC(CNC(CN)=O)=O)=O)NC([C@@H](NC([C@@H](NC([C@](N1)([H])CC3=CC=CC=C3)=O)CCC(N)=O)=O)CC(N)=O)=O)N(CCC4)[C@@H]4C(N[C@@H](CCCCN)C(NCC(N)=O)=O)=O |
| Molecular Weight (g/mol) | 1227.37 |
| Formula Weight | 1227.37 |
| Grade | Research |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.