missing translation for 'onlineSavingsMsg'
Learn More

Terbufoxon Sulfoxide, TRC

Product Code. 30008269
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008269 1 mg 1mg
30135108 10 mg 10mg
30138543 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008269 Supplier Toronto Research Chemicals Supplier No. TRCT1090851MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56165-57-2
Formula Weight 288.0619
InChI Formula InChI=1S/C9H21O4PS2/c1-6-12-14(10,13-7-2)15-8-16(11)9(3,4)5/h6-8H2,1-5H3
IUPAC Name 2-(diethoxyphosphorylsulfanylmethylsulfinyl)-2-methylpropane
Molecular Formula C9 H21 O4 P S2
Molecular Weight (g/mol) 288.36
SMILES CCOP(=O)(OCC)SCS(=O)C(C)(C)C
Synonym Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfinyl)methyl phosphorothioate, Terbufos oxon sulfoxide, Terbufoxon sulfoxide
Chemical Name or Material Terbufos-oxon-sulfoxide

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.