Learn More
TEP 7 Model Buffer Solution pH 7, WTW™
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
Description
- pH buffers by WTW that are precise and traceable to PTB/NIST in a bulk size of 1 liter with built-in dosing vessel standard buffer
- Life up to 3 years (unppened)
- Thrifty use
- Easiest handling by means of dosing device
- All modern technical buffering solutions are traceable to PTB (Physical Technical Government Agency) or NIST (National Institute of Standards and Technology, USA)
- Have accuracy that is ± 0,03 pH
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
| pH | 7 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Packaging | Bottle |
| Traceability to NIST | Yes |
Product Content Correction
Product Title
TEP 7 Model Buffer Solution pH 7, WTW™ >