Learn More
Tenuazonsäure-(acetyl-13C2)-Lösung, Honeywell Fluka™
Product Code. 15961151 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mg
Product Code. Quantity unitSize
15961151 2 mL 2mg
1 options
Product Code. 15961151
Brand: Honeywell Fluka R13232ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
50 μg/mL in acetonitrile, analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 75-05-8, 1486471-66-2
Density 0.78 g/cm3
Flash Point 6°C
Physical Form Liquid
Boiling Point 82°C
Industry Type Mycotoxins
Quantity 2 mL
Grade Analytical Standard