Learn More
Tenovin 6 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Tenovin 6 hydrochloride |
| CAS | 1011557-82-6 |
| Molecular Formula | C25 H34 N4 O2 S |
| Quantity | 25 mg |
| Synonym | N-[[[4-[[5-(Dimethylamino)-1-oxopentyl]amino]phenyl]amino]thioxomethyl]-4-(1,1-dimethylethyl)-benzamide Hydrochloride |
| InChI Formula | InChI=1S/C25H34N4O2S/c1-25(2,3)19-11-9-18(10-12-19)23(31)28-24(32)27-21-15-13-20(14-16-21)26-22(30)8-6-7-17-29(4)5/h9-16H,6-8,17H2,1-5H3,(H,26,30)(H2,27,28,31,32) |
| SMILES | CN(C)CCCCC(=O)Nc1ccc(NC(=S)NC(=O)c2ccc(cc2)C(C)(C)C)cc1 |
| IUPAC Name | 4-tert-butyl-N-[[4-[5-(dimethylamino)pentanoylamino]phenyl]carbamothioyl]benzamide |
| Molecular Weight (g/mol) | 454.63 |
| Formula Weight | 454.2402 |