missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir, MedChemExpress

Product Code. p-200097618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16446356 100 mg 100mg
16445908 10 mg 10mg
16432057 50 mg 50mg
16418547 5 mg 5mg
4 options
This item is not returnable. View return policy

Product Code. 16446356

Brand: Medchem Express HY13910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tenofovir (GS 1278) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV).

Tenofovir (GS 1278) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV).

Specifications

Chemical Name or Material Tenofovir
CAS 147127-20-6
Color White
Purity Grade Notes Research
Molecular Formula C9H14N5O4P
Quantity 100 mg
Synonym GS 1278 PMPA
Solubility Information H2O : 2 mg/mL (6.96 mM; Need ultrasonic)
SMILES C[C@@H](OCP(O)(O)=O)CN1C=NC2=C(N)N=CN=C12
Molecular Weight (g/mol) 287.21
Formula Weight 287.21
Percent Purity 98.4%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.