missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Dibenzyloxy Isopropyl Carbamate, TRC

Product Code. 30426391
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
Product Code. Quantity unitSize
30426391 25 mg -
30412818 2.5 mg 2.5mg
2 options
This item is not returnable. View return policy

Product Code. 30426391

Brand: Toronto Research Chemicals TRCT01854525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391052-09-7
Formula Weight 553.209
InChI Formula InChI=1S/C27H32N5O6P/c1-20(2)38-27(33)31-25-24-26(29-17-28-25)32(18-30-24)14-21(3)35-19-39(34,36-15-22-10-6-4-7-11-22)37-16-23-12-8-5-9-13-23/h4-13,17-18,20-21H,14-16,19H2,1-3H3,(H,28,29,31,33)/t21-/m1/s1
IUPAC Name propan-2-yl N-[9-[(2R)-2-[bis(phenylmethoxy)phosphorylmethoxy]propyl]purin-6-yl]carbamate
Molecular Formula C27H32N5O6P
Molecular Weight (g/mol) 553.55
SMILES CC(C)OC(=O)Nc1ncnc2c1ncn2C[C@@H](C)OCP(=O)(OCc3ccccc3)OCc4ccccc4
Synonym [[(1R)-2(6-Isopropylaminocarbamate-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid Dibenzyl Ester
Chemical Name or Material Tenofovir Dibenzyloxy Isopropyl Carbamate

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.