missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir alafenamide hemifumarate, MedChemExpress

Product Code. p-200097985
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16498967 5 mg 5mg
16412497 50 mg 50mg
16406686 100 mg 100mg
16406388 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16498967

Brand: Medchem Express HY15232B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tenofovir alafenamide hemifumarate (GS-7340 hemifumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.

Tenofovir alafenamide hemifumarate (GS-7340 hemifumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.

Specifications

Chemical Name or Material Tenofovir alafenamide hemifumarate
CAS 1392275-56-7
Color White
Purity Grade Notes Research
Molecular Formula C21H29N6O5P.1/2 C4H4O4
Quantity 5 mg
Synonym GS-7340 hemifumarate
Solubility Information DMSO : 50 mg/mL (93.55 mM; Need ultrasonic) ∣H2O : 20 mg/mL (37.42 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)/C=C/C(O)=O.NC1=NC=NC2=C1N=CN2C[C@@H](C)OC[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O.[0.5]
Molecular Weight (g/mol) 534.5
Formula Weight 534.5
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.