missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TENM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18036491
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18036491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18036491 Supplier Invitrogen™ Supplier No. PA5144912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DGRKPRQSYN SRETLHEYNQ ELRMNYNSQS RKRKEVEKST QEMEFCETSH TLCSGYQTDM HSVSRHGYQL EMGSDVD Antigen sequence identity: Mouse 91%; Rat 29%.

The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TENM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TENM1
Gene Accession No. Q9UKZ4
Gene Alias IDten-1; odz (odd Oz/ten-m, Drosophila) homolog 3; odz, odd Oz/ten-m homolog 1; odz, odd Oz/ten-m homolog 1(Drosophila); Odz1; Odz2; ODZ3; Protein Odd Oz/ten-m homolog 1; RGD1561678; TCAP-1; TCPA-1; Ten-1; Ten-1 ECD; Ten-1 extracellular domain; Ten-1 intracellular domain; tenascin M1; tenascin-M1; teneurin 1; Teneurin C-terminal-associated peptide; Teneurin transmembrane protein 1; teneurin-1; TENM1; Ten-m1; TNM; TNM1
Gene Symbols TENM1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 32-108. Recombinant protein control fragment (Product #RP-110036).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.