Learn More
Bochem™ Tempered Cast Iron Bossheads
Click to view available options
Diameter (Metric) Outer:
16.5 mm
36 mm
Product Type:
Bosshead
Cross Bosshead
Swivel Bosshead
Unit Size:
Each
Description
- Malleable cast iron (DIN 12895)
- Type 1: Thumb screw M8
- Type 4: Safety screw type heart M8
Specifications
Specifications
| Material | Tempered Cast Iron |
| Diameter (Metric) Outer | 16.5 mm |
| Product Type | Swivel Bosshead |
| Thread Size | M8 |
| Description | Screw Type: Swivel |
| Quantity | 1 Ea. |