Learn More
Bochem™ Tempered Cast Iron Bossheads
Product Code. 10354171
Change view
Click to view available options
Diameter (Metric) Outer:
16.5 mm
36 mm
Product Type:
Bosshead
Cross Bosshead
Swivel Bosshead
Unit Size:
Each
Product Code. Diameter (Metric) Outer Product Type unitSize
10354171 16.5 mm Bosshead Each
10120811 16.5 mm Cross Bosshead Each
10604701 16.5 mm Swivel Bosshead Each
10271632 36 mm Bosshead Each
4 options
Product Code. 10354171
Brand: Bochem™ 5360
Only null left
Add to basket
Add to basket
Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.
  • Malleable cast iron (DIN 12895)
  • Type 1: Thumb screw M8
  • Type 4: Safety screw type heart M8
Specifications
Material Tempered Cast Iron
Diameter (Metric) Outer 16.5 mm
Product Type Bosshead
Thread Size M8
Description Screw Type: 1 Thumb screw M8
Quantity 1 Ea.