Learn More
Bochem™ Tempered Cast Iron Bossheads
Product Code. 10271632
Click to view available options
Diameter (Metric) Outer:
16.5 mm
36 mm
Product Type:
Bosshead
Cross Bosshead
Swivel Bosshead
Unit Size:
Each
Product Code. 10271632
Brand: Bochem™ 5369
Only null left
Add to basket
Add to basket
Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.
  • Malleable cast iron (DIN 12895)
  • Type 1: Thumb screw M8
  • Type 4: Safety screw type heart M8
Specifications
Material Tempered Cast Iron
Diameter (Metric) Outer 36 mm
Product Type Bosshead
Thread Size M8
Description Screw Type: 1 Thumb screw M8
Quantity 1 Ea.