Description
- Additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin)
Order Info
Includes
Specifications
Specifications
| Description | Pt 100 temperature sensor - additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin) |
| For Use With (Equipment) | BINDER Chambers BD 23, BD 400 |
| Type | Temperature Sensor |
| Includes | LEMO Connector (3-Pin) |