Learn More
Thermographic™ Temperature Measurement Point Label
Provide an indication of the present temperature with a Thermographic™ Temperature Measurement Point Label. The temperature label can vary as needed to monitor and display the current temperature for sterilization.
Brand:  Thermographic™ M01ETHE099D01P1
Product Code. 11700929
  • ONLINE EXCLUSIVE PRICE
    256.00 NOK
    150.00 NOK / Pack of 50
    Save 106.00 NOK
    41% Off
Only null left
Estimated Shipment: 17-03-2026
Description
  • Heat-sensitive label with variety of temperature points
  • Adhesive
  • Irreversible