Learn More
Thermo Scientific™ Temperature Chart Recorders
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
-140°C and -150°C Freezers
Temperature Range:
-100°C to 0°C
Type:
Freestanding
Unit Size:
Each
Description
Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.
Features:
- Seven-day chart recorders record temperatures from -100° to 0°C (-40° to +38° on 6183-7)
- 6 in. (15.2cm) chart
- Use pressure-sensitive paper, require no inking
- Freestanding model is customer installed and must be installed by a qualified professional
- Panel-mounted model is factory installed; order when purchasing refrigerator or freezer
Ordering Information:
Available in range of voltages; specify voltage when ordering.
Includes:
Pack of 50 charts
Specifications
Specifications
| Description | Seven-day chart recorders |
| Type | Freestanding |
| Temperature Range | -100°C to 0°C |
| For Use With (Equipment) | -140°C and -150°C Freezers |
| Static Holding Time | 7 Days |
| Diameter (English) | 6 in. |
| Diameter (Metric) | 15.2 cm |
| Monitoring Options | Recorder |