Learn More
Thermo Scientific™ Temp 10 Series Temperature Meter, Type J
The temperature meters are available for your basic temperature measurement needs.
Brand: Thermo Scientific™ 01X450405
3296.32 NOK valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Additional Details : Weight : 0.52110kg
Description
Rugged and easy to use, the Thermo Scientific Temp 10 Series features a large, backlit dual-line LCD, giving you clear, accurate readings of measured and min/max temperature, even in dark environments. Ergonomically designed to fit your palm perfectly, the Temp 10 series offers maximum hands-free convenience with our exclusive three-way hands-free kit, which is sold separately.
Features:
User-Friendly
- Automatic calibration – just leave electrode in ice water and press ‘CAL’ to calibrate
- Min/max temperature at the press of a key
- Dual-display shows current and min/max reading
- Large, easy-to-read illuminated display
Versatile
- Measures from -250 to 1372 °C
- Accepts a wide variety of temperature probes using standard mini-connector
- Exclusive three-way hands-free option (kit sold separately) – hang the meter from a pipe or belt, stick the meter to a metallic surface, or simply stand the meter on a table surfac
Sturdy Design
- IP54 splashproof with sealed keypad and ABS plastic housing
- Protective rubber armor ensures a firm grip and doubles up as a table-top stand
Storage: -40 °C to 65 °C (-40 °F to 149 °F); 10 % to 90 % (non-condensing)
Markets
- Chemicals
- Manufacturing
- Food and Beverage
- Industrial and Automotive
Specifications
750+ hr. (without backlight) | |
Thermo Scientific type J thermocouple thermometer with rubber armor/stand (probe not included) | |
IP 54 | |
-210°C to +1200°C | |
Single (3-pin circular connector (switchcraft TA3F)) | |
-346°F to +2192°F | |
175 x 97 x 42 mm (without armor) ; 180 x 102 x 52 mm (with armor) |
3 AA 1.5 V Alkaline | |
LCD 58 x 40 mm, Backlit | |
Temperature | |
with Min/Max Reading, Hold Function, Stability Indicator, Auto Shut-Off: 17.5 min (selectable) | |
-200.0°C to -100.0°C (-392°F to -148°F): ±2.0°C (±4.0°F), -99.9°C to 199.9°C (-148°F to 392°F): ±0.2°C (±0.4°F), 200.0°C to 850.0°C (392°F to 1562°F): ±2.0°C (±4.0°F) | |
Below 1000 °: 0.1 °C/°F ; above 1000 °: 1 °C/°F | |
267 g (Without armor) ; 362 g (With armor) |