Learn More
TEM5/GPR124 Antibody (499716) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8896J
Product Code. 18582259
  • 6130.00 NOK / 0.10mL
Description
TEM5/GPR124 Monoclonal antibody specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TEM5/GPR124
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
499716
Flow Cytometry, CyTOF-ready
DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124, Met1-Leu699, Accession # Q96PE1
0.1 mL
Cancer, Endothelial Cell Markers, GPCR
25960
Store at 4°C in the dark.
IgG2b