Learn More
Promed Bioscience Telocollagen, Porcine Type I (Acidic extraction)
Lyophilized, highly purified Type I collagen (telocollagen), extracted via acid-based method, suitable for 3D cell cultures and tissue engineering applications
Brand:  Promed Bioscience PRO-2002-20
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 17502845
  • 1998.99 NOK / 20mg
Estimated Shipment: 22-05-2024
Description
  • Lyophilized, highly purified Type I collagen (telocollagen), extracted via acid-based method
  • Telocollagen retains the fibrillar structure of the native molecule and preserves the superior physical and mechanical properties of the collagen found in tissues
  • Suitable as a coating for cell culture labware, for 2D and 3D cell cultures (e.g. higher stiffness scaffolds) and for tissue engineering applications
Specifications
Specifications
Lyophilized Fibrous Powder
20 mg
<1.0 EU/mL
Dialysis
-20°C to 8°C
>95% (by SDS/Western blotting)