missing translation for 'onlineSavingsMsg'
Learn More

TELO2 Mouse anti-Human, Rat, Clone: 3H10F4, Proteintech

Product Code. 16857625 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857625 20 μL 20µL
16847625 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857625

Brand: Proteintech 660771IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.

Specifications

Antigen TELO2
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 3H10F4
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TELO2
Gene Accession No. Q9Y4R8
Gene Alias hCLK2, KIAA0683, Protein clk 2 homolog, TEL2, TELO2
Gene Symbols Telo2
Host Species Mouse
Immunogen TELO2 Fusion Protein Ag8834
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302986, 9894
Target Species Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.