Learn More
Corning™ Telescopic Suction Tubes
Product Code. 16862110 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
25 to 100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
16862110 2.5 to 10 mL Each
16872110 25 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16862110 Supplier Corning™ Supplier No. 6867
  • ONLINE EXCLUSIVE PRICE
    312.00 NOK
    291.35 NOK / Pack of 1
    Save 20.65 NOK
    6% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
Corning™ Telescopic Suction Tubes for quick and precise dispensing of acids, bases and solvents
  • Suction tube
  • For use with Bottle Top Dispenser
TRUSTED_SUSTAINABILITY
Specifications
Product Type Telescopic Suction Tube
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Volume (Metric) 2.5 to 10 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) 2.5-10 mL Bottle Top Dispenser