Learn More
Heidolph™ Telescope Stand
Product Code. 10682614 Shop All Heidolph Instruments Products
Change view
Click to view available options
Diameter (Metric) Rod:
32 mm
Unit Size:
Each
Product Code. Diameter (Metric) Rod unitSize
10682614 32 mm Each
1 options
Product Code. 10682614
Brand: Heidolph™ 5701210000
Ideal Telescope Stand for easy mounting of overhead stirrers.
  • Stand Heidolph Telescopic
  • Universal Stand S2
  • Adjustable length 725 mm to 1025 mm
  • Tube diameter 32 mm
TRUSTED_SUSTAINABILITY
Specifications
Length (English) Rod 28.5 to 40.4 in.
Length (Metric) Rod 725 to 1025 mm
Diameter (English) Rod 1.3 in.
Diameter (Metric) Rod 32 mm
Weight (English) 17 lb.
Weight (Metric) 7.7 kg