missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12959-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.Specifications
| TEKT3 | |
| Polyclonal | |
| Unconjugated | |
| TEKT3 | |
| TEKT3, tektin 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287392, 64518, 71062 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V8G8, Q6X6Z7, Q9BXF9 | |
| Tekt3 | |
| TEKT3 Fusion Protein Ag4080 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction