missing translation for 'onlineSavingsMsg'
Learn More

TEKT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12959-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850944

  • 1415.13 NOK / 20µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
Specifications

Specifications

TEKT3
Polyclonal
Unconjugated
TEKT3
TEKT3, tektin 3
Rabbit
Antigen Affinity Chromatography
RUO
287392, 64518, 71062
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4V8G8, Q6X6Z7, Q9BXF9
Tekt3
TEKT3 Fusion Protein Ag4080
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers