Learn More
TEKT2 Antibody (OTI1E6), Novus Biologicals Biologicals™
Product Code. 30306711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
30306711 30 μL 30µL
30306298 100 μL 100µL
2 options
Product Code. 30306711
Brand: Novus Biologicals NBP34504230ul
Mouse Monoclonal Antibody
TEKT2 Monoclonal antibody specifically detects TEKT2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen TEKT2
Applications Western Blot
Classification Monoclonal
Clone OTI1E6
Conjugate Unconjugated
Dilution Western Blot 1:500-2000
Formulation PBS (pH 7.3) 1% BSA, 50% glycerol
Gene Alias h-tektin-t, TEKTB1tektin-B1, tektin 2 (testicular), tektin-2, Tektin-B1, TEKTIN-T, Testicular tektin, Testicular tektin B1-like protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-244 of human TEKT2(NP_055281) produced in E.coli.
Purification Method >95%
Quantity 30 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27285
Target Species Human
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1