missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18968-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predomitly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.Specifications
TEKT1 | |
Polyclonal | |
Unconjugated | |
TEKT1 | |
TEKT1, tektin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21689, 83659, 85270 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q969V4, Q99JD2, Q9DAJ2 | |
Tekt1 | |
TEKT1 Fusion Protein Ag5173 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |