missing translation for 'onlineSavingsMsg'
Learn More

TEKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18968-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898764

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predomitly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.
Specifications

Specifications

TEKT1
Polyclonal
Unconjugated
TEKT1
TEKT1, tektin 1
Rabbit
Antigen Affinity Chromatography
RUO
21689, 83659, 85270
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969V4, Q99JD2, Q9DAJ2
Tekt1
TEKT1 Fusion Protein Ag5173
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers