missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ Tegrant ThermoSafe LabMailer Absorbent Sheets

Product Code. 13501930
Change view
Click to view available options
Capacity:
22.5 mL of blood/sheet
Diameter (Metric) Interior:
7.6 x 7.6 cm
Diameter (English) Interior:
3 x 3 in.
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Diameter (Metric) Interior Diameter (English) Interior Unit Size
13501930 22.5 mL of blood/sheet 7.6 x 7.6 cm 3 x 3 in. Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13501930 Supplier Sonoco™ Supplier No. Zorb33

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lightweight, economical lab mailer shippers

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Polystyrene foam construction saves postage
  • Reusable and recyclable

Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 22.5mL of blood per sheet
For Use With (Equipment) Provide maximum breakage protection for known non-hazardous materials.
Applications For protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
Diameter (English) Interior 3 x 3 in.
Diameter (Metric) Interior 7.6 x 7.6 cm
Capacity 22.5 mL of blood/sheet
Material Lightweight Polystyrene Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.