Learn More
RETSCH Teflon Grinding Jars
Brand: RETSCH 02.462.0051
5875.75 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.07000kg
Specifications
Grinding Jar | |
For MM 200 | |
Teflon |
25 mL | |
20 mm |