Learn More
Technical Buffer for pH DKD Certified pH 7.00, METTLER TOLEDO™
GREENER_CHOICE
Product Code. p-8014306 Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Packaging:
1Each
6x 250 mL
Unit Size:
1500mL
250mL
Product Code. 10709883
Brand: Mettler Toledo 51350034
The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.
  • pH 7.00
TRUSTED_SUSTAINABILITY
Specifications
pH 7.00
Quantity 250 mL
Packaging 1Each