Learn More
Cole-Parmer™ Essentials Cell Culturing Flask for BS-200 Series Cell Culture Stirring Systems
Product Code. 11583779
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 11583779
Brand: Cole-Parmer™ Essentials 2183737
Only null left
Add to basket
Add to basket
This Cell Culturing Flask is a complete culture vessel—flask, stir rod, and caps—to use with biological stirrers.
  • Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.
Specifications
Capacity (Metric) 125 mL
Material Borosilicate Glass
Height (English) 4.9 in.
Height (Metric) 125 mm
Autoclavable Autoclavable
Diameter (Metric) Outer 65 mm