Learn More
Techne™ PrimePro 48 Real-Time PCR System Accessories
Product Code. 15303165
Change view
Click to view available options
No. of Wells:
48
Unit Size:
Each
Product Code. No. of Wells unitSize
15303165 48 Each
1 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 15303165
Brand: Techne™ 9394598
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Techne™ PrimePro 48 Real-Time PCR System Accessories keep you running smoothly.
Plates and seals for use with your Techne PrimePro 48 Thermal Cycler
Specifications
Format Standard
Description Real-Time PCR Plates for Prime Pro 48; 50/Pk
Array 6 x 8
Color Natural Translucent
For Use With (Equipment) Techne PrimePro 48
Material Polypropylene
No. of Wells 48
No. per Pack 50