Learn More
TEC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18085-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16848124
  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
Description
The protein encoded by this gene belongs to the Tec family of non-receptor protein-tyrosine kinases containing a pleckstrin homology domain. Tec family kinases are involved in the intracellular signaling mechanisms of cytokine receptors, lymphocyte surface antigens, heterotrimeric G-protein coupled receptors, and integrin molecules. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
Specifications
Specifications
TEC
Polyclonal
Unconjugated
TEC
PSCTK4, TEC, tec protein tyrosine kinase, Tyrosine protein kinase Tec
Rabbit
Antigen Affinity Chromatography
RUO
100124696, 21682, 84492
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P24604, P42680
Tec
TEC Fusion Protein Ag12804
150 μL
Primary
Human, Mouse, Rat