Learn More
Tebuconazole, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Tebuconazole |
| CAS | 107534-96-3 |
| Analyte or Component Names | Tebuconazole |
| Shipping Condition | +20°C |
| Molecular Formula | C16H22ClN3O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1H-1,2,4-Triazole-1-ethanol, α-[2-(4-chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-, (±)-, α-[2-(4-Chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol, (±)-Tebuconazole, Atlas, BAY-HWG 1608, Corail, Dedalus 25 WDG, Elite, Elite (fungicide), Ethyltrianol, Etiltrianol, Fenetrazole, Folicur, Folicur 200EC, Folicur 3.6F, Gelseal, Gelseal (fungicide), Greenseal, HWG 1608, Hao Li-ke, Horizon, Horizon (fungicide), Horizont, Microban S 2142, Microban TZ, ONLY ONE Flowable, Orius, Orius (fungicide), Orius 25EC, Preventol A 8, Raxil, Raxil 2DS, Raxil Ultra, Rival 200EC, Riza, Rosacur, S 2142, Silvacure, Sparta, Sparta 250EW, Tebu 250EW, Tebu 60, Tebuconazole, Terbutrazole, Timbertreat T;Formerly 80443-41-0 |
| InChI Formula | InChI=1S/C16H22ClN3O/c1-15(2,3)16(21,10-20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3 |
| SMILES | CC(C)(C)C(O)(CCc1ccc(Cl)cc1)Cn2cncn2 |
| IUPAC Name | 1-(4-chlorophenyl)-4,4-dimethyl-3-(1,2,4-triazol-1-ylmethyl)pentan-3-ol |
| Show More |