Learn More
TEAD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21159-1-AP-20UL
 View more versions of this product
Product Code. 16810775
  • 1710.33 NOK / 20µL
Description
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5′-GTGGAATGT-3′). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.
Specifications
Specifications
TEAD2
Polyclonal
Unconjugated
TEAD2
ETF, TEA domain family member 2, TEAD 2, TEAD2, TEF 4, TEF4
Rabbit
Antigen Affinity Chromatography
RUO
21677, 8463
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48301, Q15562
TEAD2
TEAD2 Fusion Protein Ag14074
20 μL
Primary
Mouse, Human